4-Carboxy-1-naphthaleneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Borono-1-naphthoic acid

A560979-100MG A560979-1G A560979-250MG
77292-526EA 35.02 USD
77292-526 77292-528 77292-530
4-Carboxy-1-naphthaleneboronic acid 98%
4-Carboxy-1-naphthaleneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₉BO₄
MW: 216.00 g/mol
MDL Number: MFCD06203538
CAS Number: 332398-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR