2-Amino-3,5-dimethylpyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethylpyrazin-2-amine

A560818-1G A560818-250MG A560818-25G A560818-5G
77280-034EA 44.29 USD
77280-034 77280-036 77280-038 77280-040
2-Amino-3,5-dimethylpyrazine 98%
2-Amino-3,5-dimethylpyrazine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MW: 123.1558 g/mol
MDL Number: MFCD00107172
CAS Number: 91678-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR