Methyl 4-cyanofuran-2-carboxylate 95%

Supplier: Ambeed
Warning

A560696-100MG A560696-1G A560696-50MG
77266-344EA 59 USD
77266-344 77266-346 77266-348
Methyl 4-cyanofuran-2-carboxylate 95%
Methyl 4-cyanofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅NO₃
MDL Number: MFCD16658901
CAS Number: 357289-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR