Monosodium 2-sulfoterephthalate 98%

Supplier: Ambeed
Warning

Synonyms: Monosodium 2-sulphoterephthalate, 2-Sulphoterephthalic acid Monosodium salt

A560615-25G A560615-5G A560615-1G
77242-050EA 148 USD
77242-050 77242-052 77594-696
Monosodium 2-sulfoterephthalate 98%
Monosodium 2-sulfoterephthalate

Precaución: for Research Use Only

Formula: C₈H₆O₇S
MW: 268.17 g/mol
MDL Number: MFCD00059843
CAS Number: 19089-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR