tert-Butyl 6-bromohexanoate 95%

Supplier: Ambeed
Warning

A560585-100MG A560585-1G A560585-250MG A560585-5G A560585-25G A560585-100G
77253-958EA 5 USD
77253-958 77253-960 77253-962 77253-964 77551-682 77747-155
tert-Butyl 6-bromohexanoate 95%
tert-Butyl 6-bromohexanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉BrO₂
MDL Number: MFCD00457120
CAS Number: 65868-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR