2-((2-Methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-11-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one 97%
Supplier: Ambeed
|
Warning
|
A560552-10MG
A560552-1MG
A560552-25MG
A560552-5MG
77266-336EA
97
USD
77266-336
77266-338
77266-340
77266-342
2-((2-Methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-11-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one 97%
2-((2-Methoxy-4-(4-methylpiperazin-1-yl)phenyl)amino)-11-methyl-5,11-dihydro-6H-benzo[e]pyrimido[5,4-b][1,4]diazepin-6-one
Precaución: for Research Use Only
Formula:
C₂₄H₂₇N₇O₂ MW: 445.52 g/mol |
MDL Number:
MFCD30742941 CAS Number: 1234480-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...