2,6-Dimethoxy-p-cresol 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol

A560448-1G A560448-25G A560448-5G A560448-100G
77253-952EA 12.36 USD
77253-952 77253-954 77253-956 77747-154
2,6-Dimethoxy-p-cresol 95%
2,6-Dimethoxy-p-cresol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
MDL Number: MFCD00017289
CAS Number: 6638-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR