4-(5-Oxazolyl)benzonitrile 97%

Supplier: Ambeed
Warning

A560326-1G A560326-250MG A560326-5G
77292-520EA 52 USD
77292-520 77292-522 77292-524
4-(5-Oxazolyl)benzonitrile 97%
4-(5-Oxazolyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O
MDL Number: MFCD00084987
CAS Number: 87150-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR