2-Ethoxy-5-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(2-ethoxy-5-fluorophenyl)borane, (2-Ethoxy-5-fluorophenyl)boronic acid, 2-Ethoxy-5-fluorobenzeneboronic acid, (2-Ethoxy-5-fluorophenyl)metaboric acid

A559894-100G A559894-1G A559894-25G A559894-5G
77280-024EA 471.74 USD
77280-024 77280-026 77280-028 77280-030
2-Ethoxy-5-fluorophenylboronic acid 98%
2-Ethoxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD05664328
CAS Number: 864301-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR