BQU57 98+%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-1,3-dimethyl-4-(4-(trifluoromethyl)phenyl)-1,4-dihydropyrano[2,3-c]pyrazole-5-carbonitrile

A559382-10MG A559382-1MG A559382-50MG A559382-5MG
77242-042EA 39.14 USD
77242-042 77242-044 77242-046 77242-048
BQU57 98+%
BQU57

Precaución: for Research Use Only

Formula: C₁₆H₁₃F₃N₄O
MW: 334.3 g/mol
MDL Number: MFCD28902198
CAS Number: 1637739-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR