4,4'-(Propane-2,2-diyl)bis((allyloxy)benzene) 97%

Supplier: Ambeed
Warning

A558974-100G A558974-25G A558974-500G
77292-500EA 24.72 USD
77292-500 77292-502 77292-504
4,4'-(Propane-2,2-diyl)bis((allyloxy)benzene) 97%
4,4'-(Propane-2,2-diyl)bis((allyloxy)benzene)

Precaución: for Research Use Only

Formula: C₂₁H₂₄O₂
MW: 308.41 g/mol
MDL Number: MFCD01218874
CAS Number: 3739-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR