1-Hydroxy-4-nitro-9,10-anthraquinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-4-nitroanthracene-9,10-dione

A558939-100MG A558939-1G A558939-250MG
77242-036EA 8.24 USD
77242-036 77242-038 77242-040
1-Hydroxy-4-nitro-9,10-anthraquinone 98%
1-Hydroxy-4-nitro-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₇NO₅
MW: 269.2091 g/mol
MDL Number: MFCD00142642
CAS Number: 81-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR