3,5-Dibromo-p-cresol 95%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dibromo-5-hydroxy-2-methylbenzene, 2,6-Dibromo-4-hydroxytoluene, 3,5-Dibromo-4-methylphenol

A558471-1G A558471-250MG A558471-5G
77280-018EA 64.89 USD
77280-018 77280-020 77280-022
3,5-Dibromo-p-cresol 95%
3,5-Dibromo-p-cresol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MW: 265.93 g/mol
MDL Number: MFCD00013968
CAS Number: 13979-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR