2,2',2''-(Benzene-1,3,5-triyl)triacetonitrile 98%

Supplier: Ambeed
Danger

A558303-1G A558303-250MG
77242-022EA 435.69 USD
77242-022 77242-024
2,2',2''-(Benzene-1,3,5-triyl)triacetonitrile 98%
2,2',2''-(Benzene-1,3,5-triyl)triacetonitrile

Precaución: for Research Use Only

Formula: C₁₂H₉N₃
MW: 195.22 g/mol
MDL Number: MFCD30491308
CAS Number: 80935-59-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR