4-Chlorochalcone 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Chlorophenyl)-1-phenylprop-2-en-1-one, 4-Chlorobenzylidenacetophenone

A558202-25G A558202-5G
77253-926EA 77.25 USD
77253-926 77253-928
4-Chlorochalcone 98%
4-Chlorochalcone

Precaución: for Research Use Only

Formula: C₁₅H₁₁ClO
MW: 242.7 g/mol
MDL Number: MFCD00016345
CAS Number: 956-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR