3-(2,3-Dihydroindol-1-yl)propylamine 95%

Supplier: Ambeed
Warning

A558007-100MG A558007-1G A558007-250MG
77280-012EA 45 USD
77280-012 77280-014 77280-016
3-(2,3-Dihydroindol-1-yl)propylamine 95%
3-(2,3-Dihydroindol-1-yl)propylamine

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂
MDL Number: MFCD03011732
CAS Number: 61123-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR