4,4'-Dichloro-2,2'-bipyridine 96%

Supplier: Ambeed
Danger

A557959-1G A557959-250MG A557959-5G
77292-494EA 72 USD
77292-494 77292-496 77749-152
4,4'-Dichloro-2,2'-bipyridine 96%
4,4'-Dichloro-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₆Cl₂N₂
MDL Number: MFCD00233879
CAS Number: 1762-41-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR