3-Nitroimidazo[1,2-b]pyridazine 95%

Supplier: Ambeed
Warning

A557556-100MG A557556-1G A557556-250MG A557556-5G
77292-480EA 40.17 USD
77292-480 77292-482 77292-484 77292-486
3-Nitroimidazo[1,2-b]pyridazine 95%
3-Nitroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₄N₄O₂
MW: 164.12 g/mol
MDL Number: MFCD09832519
CAS Number: 18087-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR