1-(((2R,3S)-2-(2,5-Difluorophenyl)-3-methyloxiran-2-yl)methyl)-1H-1,2,4-triazole 97%

Supplier: Ambeed
Warning

A557258-1G A557258-25G
77242-014EA 161.71 USD
77242-014 77242-016
1-(((2R,3S)-2-(2,5-Difluorophenyl)-3-methyloxiran-2-yl)methyl)-1H-1,2,4-triazole 97%
1-(((2R,3S)-2-(2,5-Difluorophenyl)-3-methyloxiran-2-yl)methyl)-1H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₂N₃O
MW: 251.23 g/mol
CAS Number: 241479-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR