(2,3,5-Trifluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A557250-100MG A557250-1G A557250-250MG A557250-5G
77242-006EA 11 USD
77242-006 77242-008 77242-010 77242-012
(2,3,5-Trifluorophenyl)acetic acid 98%
(2,3,5-Trifluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₂F₃CH₂COOH
MDL Number: MFCD00083547
CAS Number: 132992-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR