PNU-74654 98%

Supplier: Ambeed
Warning

Synonyms: N'-((5-Methylfuran-2-yl)methylene)-2-phenoxybenzohydrazide

A557216-100MG A557216-10MG A557216-1MG A557216-25MG A557216-50MG A557216-5MG
77266-306EA 345.05 USD
77266-306 77266-308 77266-310 77266-312 77266-314 77266-316
PNU-74654 98%
PNU-74654

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₂O₃
MW: 320.3419 g/mol
MDL Number: MFCD12912438
CAS Number: 113906-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR