(6-(Difluoromethoxy)pyridin-3-yl)methanol 97%

Supplier: Ambeed
Warning

A557123-100MG A557123-250MG
77266-292EA 52.53 USD
77266-292 77266-294
(6-(Difluoromethoxy)pyridin-3-yl)methanol 97%
(6-(Difluoromethoxy)pyridin-3-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₇F₂NO₂
MW: 175.13 g/mol
MDL Number: MFCD28339841
CAS Number: 1375098-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR