Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride

A557073-1000G A557073-100G A557073-25G A557073-500G
77266-284EA 80.34 USD
77266-284 77266-286 77266-288 77266-290
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) 98%
Girard's reagent T (carbazoylmethyltrimethylammonium chloride)

Precaución: for Research Use Only

Formula: C₅H₁₄ClN₃O
MW: 167.64 g/mol
MDL Number: MFCD00012009
CAS Number: 123-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR