(3R,4R)-tert-Butyl 3-amino-4-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A557044-100MG
A557044-1G
A557044-250MG
77266-278EA
143.17
USD
77266-278
77266-280
77266-282
(3R,4R)-tert-Butyl 3-amino-4-(hydroxymethyl)pyrrolidine-1-carboxylate 98%
(3R,4R)-tert-Butyl 3-amino-4-(hydroxymethyl)pyrrolidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₂₀N₂O₃ MW: 216.28 g/mol |
MDL Number:
MFCD12031285 CAS Number: 1207853-71-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...