(S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A557010-100MG
A557010-1G
A557010-250MG
77253-920EA
46.35
USD
77253-920
77253-922
77253-924
(S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(S)-5,5',6,6',7,7',8,8'-Octahydro-3,3'-diphenyl-[1,1'-binaphthalene]-2,2'-diol
Precaución: for Research Use Only
Formula:
C₃₂H₃₀O₂ MW: 446.58 g/mol |
MDL Number:
MFCD30541639 CAS Number: 575451-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...