4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran 98%

Supplier: Ambeed
Danger

A556764-1G A556764-250MG A556764-25G A556764-5G
77241-998EA 54 USD
77241-998 77242-000 77242-002 77242-004
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran 98%
4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MDL Number: MFCD00181519
CAS Number: 28286-88-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR