Ethyl-2-(hydroxymethyl)acrylate 95% stabilized
Supplier: Ambeed
|
Danger
|
Stabilization:
Stabilised with hydroquinone (HQ)
A556448-10G
A556448-1G
A556448-250MG
A556448-5G
77253-914EA
37
USD
77253-914
77253-916
77253-918
77747-145
Ethyl-2-(hydroxymethyl)acrylate 95% stabilized
Ethyl-2-(hydroxymethyl)acrylate
Precaución: for Research Use Only
Formula:
C₆H₁₀O₃ |
MDL Number:
MFCD01673859 CAS Number: 10029-04-6 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...