2,6-Diamino-4-chloropyrimidine-1-oxide 97%

Supplier: Ambeed
Warning

A556354-100MG A556354-1G A556354-250MG A556354-5G
77292-470EA 14.42 USD
77292-470 77292-472 77292-474 77749-149
2,6-Diamino-4-chloropyrimidine-1-oxide 97%
2,6-Diamino-4-chloropyrimidine-1-oxide

Precaución: for Research Use Only

Formula: C₄H₅ClN₄O
MW: 160.56 g/mol
MDL Number: MFCD00134274
CAS Number: 35139-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR