2,4,6-Tribromopyrimidine 98%

Supplier: Ambeed
Warning

A556295-1G A556295-5G
77253-910EA 55 USD
77253-910 77253-912
2,4,6-Tribromopyrimidine 98%
2,4,6-Tribromopyrimidine

Precaución: for Research Use Only

Formula: C₄HBr₃N₂
MW: 316.78
MDL Number: MFCD00233942
CAS Number: 36847-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR