Ethyl-3,5-dihydroxybenzoate 98%

Supplier: Ambeed
Warning

A556285-100G A556285-1G A556285-25G A556285-5G
77253-902EA 40 USD
77253-902 77253-904 77253-906 77253-908
Ethyl-3,5-dihydroxybenzoate 98%
Ethyl-3,5-dihydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MDL Number: MFCD00020122
CAS Number: 4142-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR