5,5',5''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))triisophthalic acid 97%

Supplier: Ambeed
Warning

A555996-100MG A555996-1G A555996-250MG A555996-5G
77266-264EA 26.78 USD
77266-264 77266-266 77266-268 77266-270
5,5',5''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))triisophthalic acid 97%
5,5',5''-((1,3,5-Triazine-2,4,6-triyl)tris(azanediyl))triisophthalic acid

Precaución: for Research Use Only

Formula: C₂₇H₁₈N₆O₁₂
MW: 618.46 g/mol
CAS Number: 776242-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR