4,6-Bis((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan ≥98%, ee 99%

Supplier: Ambeed
Warning

A554481-1G A554481-250MG A554481-100MG
77266-258EA 509.85 USD
77266-258 77266-260 77749-144
4,6-Bis((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan ≥98%, ee 99%
4,6-Bis((S)-4-Phenyl-4,5-dihydrooxazol-2-yl)dibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₃₀H₂₂N₂O₃
MW: 458.51 g/mol
MDL Number: MFCD32664427
CAS Number: 246040-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR