2,3,5-Tribromothiophene 98%

Supplier: Ambeed
Warning

A553844-100G A553844-5G
77292-450EA 70.04 USD
77292-450 77292-452
2,3,5-Tribromothiophene 98%
2,3,5-Tribromothiophene

Precaución: for Research Use Only

Formula: C₄HBr₃S
MW: 320.83 g/mol
MDL Number: MFCD00014521
CAS Number: 3141-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR