AS101 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ammonium trichloro[1,2-ethanediolato-O,O']-tellurate, AS 101
A553186-10MG
A553186-1MG
A553186-25MG
A553186-50MG
A553186-5MG
77266-248EA
178.19
USD
77266-248
77266-250
77266-252
77266-254
77266-256
AS101 98%
AS101
Precaución: for Research Use Only
Formula:
C₂H₈Cl₃NO₂Te MW: 312.04 g/mol |
MDL Number:
MFCD01682758 CAS Number: 106566-58-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...