1H-Pyrazolo[3,4-b]pyridin-5-ol 97%
Supplier: Ambeed
|
Warning
|
A553165-100MG
A553165-250MG
77266-244EA
234.84
USD
77266-244
77266-246
1H-Pyrazolo[3,4-b]pyridin-5-ol 97%
1H-Pyrazolo[3,4-b]pyridin-5-ol
Precaución: for Research Use Only
Formula:
C₆H₅N₃O MW: 135.12 g/mol |
MDL Number:
MFCD18255090 CAS Number: 1256818-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...