3,5-Dimethyl-4-nitropyridine 1-oxide 95%
Supplier: Ambeed
|
Warning
|
A552809-25G
A552809-5G
77280-002EA
94.76
USD
77280-002
77280-004
3,5-Dimethyl-4-nitropyridine 1-oxide 95%
3,5-Dimethyl-4-nitropyridine 1-oxide
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₃ MW: 168.15 g/mol |
MDL Number:
MFCD00956001 CAS Number: 14248-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...