1,3,6,8-Tetramethyl-9H-carbazole 98%

Supplier: Ambeed
Warning

A552586-250MG A552586-1G
77266-238EA 70.04 USD
77266-238 77749-143
1,3,6,8-Tetramethyl-9H-carbazole 98%
1,3,6,8-Tetramethyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₆H₁₇N
MW: 223.32 g/mol
MDL Number: MFCD18450169
CAS Number: 6558-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR