2-Bromo-1-(2-chloropyridin-4-yl)ethanone 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-Bromo-1-(2-chloro-4-pyridyl)ethanone
A552379-100MG
A552379-1G
A552379-250MG
77266-230EA
64.89
USD
77266-230
77266-232
77266-234
2-Bromo-1-(2-chloropyridin-4-yl)ethanone 95%
2-Bromo-1-(2-chloropyridin-4-yl)ethanone
Precaución: for Research Use Only
Formula:
C₇H₅BrClNO MW: 234.478 g/mol |
MDL Number:
MFCD09702373 CAS Number: 23794-16-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...