Norcantharidin 98%

Supplier: Ambeed
Warning

Synonyms: exo-7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic Anhydride, rel-(3aR,4R,7S,7aS)-Hexahydro-4,7-epoxyisobenzofuran-1,3-dione, exo-3,6-Epoxyhexahydrophthalic Anhydride

A552376-1G A552376-250MG A552376-5G A552376-25G
77253-842EA 18.54 USD
77253-842 77253-844 77253-846 77747-137
Norcantharidin 98%
Norcantharidin

Precaución: for Research Use Only

Formula: C₈H₈O₄
MW: 168.15 g/mol
MDL Number: MFCD00213361
CAS Number: 29745-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR