[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid 98%

Supplier: Ambeed
Warning

A552360-100MG A552360-250MG
77253-840EA 17 USD
77253-840 77747-136
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid 98%
[1,1':3',1''-Terphenyl]-4,4''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₄O₄
MDL Number: MFCD30543297
CAS Number: 13215-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR