MK-4101 98+%

Supplier: Ambeed
Warning

Synonyms: 5-(3,3-Difluorocyclobutyl)-3-[4-[4-methyl-5-[2-(trifluoromethyl)phenyl]-4H-1,2,4-triazol-3-yl]bicyclo[2.2.2]oct-1-yl]-1,2,4-oxadiazole

A551820-10MG A551820-1MG A551820-25MG A551820-50MG A551820-5MG
77253-830EA 63.86 USD
77253-830 77253-832 77253-834 77253-836 77253-838
MK-4101 98+%
MK-4101

Precaución: for Research Use Only

Formula: C₂₄H₂₄F₅N₅O
MW: 493.48 g/mol
CAS Number: 935273-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR