4-Fluoro-2,3-dimethylphenol 95%

Supplier: Ambeed
Warning

A551719-100MG A551719-1G A551719-250MG A551719-5G
77253-822EA 6 USD
77253-822 77253-824 77253-826 77253-828
4-Fluoro-2,3-dimethylphenol 95%
4-Fluoro-2,3-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD00153130
CAS Number: 77249-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR