Methyl 2-(3-bromo-2-fluorophenyl)acetate 95%

Supplier: Ambeed
Warning

A551453-100MG A551453-250MG A551453-1G
77266-226EA 48 USD
77266-226 77266-228 77749-142
Methyl 2-(3-bromo-2-fluorophenyl)acetate 95%
Methyl 2-(3-bromo-2-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MDL Number: MFCD22988857
CAS Number: 1427451-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR