2,1-Benzisoxazole 98%

Supplier: Ambeed
Warning

Synonyms: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

A550798-1G A550798-250MG A550798-25G
77253-816EA 28.84 USD
77253-816 77253-818 77253-820
2,1-Benzisoxazole 98%
2,1-Benzisoxazole

Precaución: for Research Use Only

Formula: C₇H₅NO
MW: 119.1207 g/mol
MDL Number: MFCD00005806
CAS Number: 271-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR