4,4'-((2,2-Bis((4-carboxyphenoxy)methyl)propane-1,3-diyl)bis(oxy))dibenzoic acid 97%

Supplier: Ambeed
Warning

A550382-100MG A550382-1G A550382-250MG A550382-5G
77266-218EA 61.8 USD
77266-218 77266-220 77266-222 77266-224
4,4'-((2,2-Bis((4-carboxyphenoxy)methyl)propane-1,3-diyl)bis(oxy))dibenzoic acid 97%
4,4'-((2,2-Bis((4-carboxyphenoxy)methyl)propane-1,3-diyl)bis(oxy))dibenzoic acid

Precaución: for Research Use Only

Formula: C₃₃H₂₈O₁₂
MW: 616.56 g/mol
CAS Number: 245551-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR