2-Aminophenyl phenyl sulfide 95%

Supplier: Ambeed
Danger

Synonyms: 2-Aminodiphenyl Sulphide, 2-(Phenylthio)aniline

A550228-100G A550228-25G A550228-5G
77266-212EA 51 USD
77266-212 77266-214 77266-216
2-Aminophenyl phenyl sulfide 95%
2-Aminophenyl phenyl sulfide

Precaución: for Research Use Only

Formula: C₁₂H₁₁NS
MW: 201.29 g/mol
MDL Number: MFCD00190139
CAS Number: 1134-94-7
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR