8-Mercaptoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: Thiooxine Hydrochloride, 8-Mercaptoquinoline Hydrochloride

A550020-1G A550020-5G
77266-208EA 59 USD
77266-208 77266-210
8-Mercaptoquinoline hydrochloride 95%
8-Mercaptoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₇NS
MDL Number: MFCD00043261
CAS Number: 34006-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR