4-(Methoxycarbonyl)nicotinic acid 98%

Supplier: Ambeed
Warning

A549887-1G A549887-250MG A549887-5G
77292-440EA 48.41 USD
77292-440 77292-442 77292-444
4-(Methoxycarbonyl)nicotinic acid 98%
4-(Methoxycarbonyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00111513
CAS Number: 24202-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR