2,4-Diamino-6-methyl-1,3,5-triazine 96%

Supplier: Ambeed
Warning

Synonyms: 2,4-Diamino-6-methyl-1,3,5-triazine 6-Methyl-1,3,5-triazine-2,4-diamine, 2,4-Diamino-6-methyl-s-triazine, 6-Methyl-1,3,5-triazine-2,4-diamine, Acetoguanamine, 6-Methyl-1,3,5-triazine-2,4-diyldiamine

A549733-1000G A549733-100G A549733-25G A549733-500G
77253-808EA 87.55 USD
77253-808 77253-810 77253-812 77253-814
2,4-Diamino-6-methyl-1,3,5-triazine 96%
2,4-Diamino-6-methyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₄H₇N₅
MW: 125.13 g/mol
MDL Number: MFCD00023192
CAS Number: 542-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR