1-Chlorotetradecane 98%

Supplier: Ambeed
Warning

A549351-100G A549351-25G A549351-500G
77241-958EA 30 USD
77241-958 77241-960 77241-962
1-Chlorotetradecane 98%
1-Chlorotetradecane

Precaución: for Research Use Only

Formula: C₁₄H₂₉Cl
MDL Number: MFCD00000958
CAS Number: 2425-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR